Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031617.2 | NP_001026788.1 | 3 | 1726 | ||
GenBank mRNA | AK023185.1 | - | 3 | 1662 | ||
AY957566.1 | - | 3 | 1676 | |||
BC110420.1 | - | 3 | 1663 | 5208612 |