Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49s3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153726.1 | NP_714948.1 | 4 | 386 | ||
GenBank mRNA | AY102036.1 | - | 4 | 386 | ||
AY747628.1 | - | 5 | 437 |