Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019613.3 | NP_062559.2 | 5, 6 | 630 | ||
XM_005256377.4 | XP_005256434.1 | 4, 5 | 453 | |||
XM_017024862.1 | XP_016880351.1 | 6, 7 | 1967 | |||
XM_017024863.1 | XP_016880352.1 | 7, 8 | 765 | |||
GenBank mRNA | AF091083.1 | - | 3, 4 | 273 | ||
AK297477.1 | - | 5, 6 | 384 | |||
AM182326.1 | - | 5, 6 | 435 | |||
AY691427.1 | - | 3, 4 | 261 | |||
BC000974.2 | - | 5, 6 | 413 | |||
BC007838.1 | - | 5, 6 | 602 | |||
CR456770.1 | - | 3, 4 | 261 |