Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025624.2 | NP_001020795.1 | 6 | 568 | ||
GenBank mRNA | AF329828.1 | - | 1 | 104 | ||
BC099178.1 | - | 6 | 534 | 7383003 | ||
BC099815.1 | - | 6 | 543 | 7929654 | ||
FQ213300.1 | - | 6 | 535 | |||
FQ213727.1 | - | 6 | 528 | |||
FQ214234.1 | - | 4 | 361 | |||
FQ214394.1 | - | 6 | 527 | |||
FQ214422.1 | - | 6 | 538 | |||
FQ215102.1 | - | 6 | 545 | |||
FQ216264.1 | - | 6 | 561 | |||
FQ216489.1 | - | 6 | 500 | |||
FQ219745.1 | - | 6 | 603 | |||
FQ219836.1 | - | 6 | 565 | |||
FQ220601.1 | - | 6 | 579 | |||
FQ221300.1 | - | 6 | 509 | |||
FQ223249.1 | - | 6 | 526 | |||
FQ229179.1 | - | 6 | 527 | |||
FQ229184.1 | - | 6 | 500 |
Gene Symbol: LOC100360316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751587.3 | XP_003751635.1 | 1 | 531 |