Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020121.3 | NP_064506.3 | 4, 5 | 663 | ||
XM_011521094.1 | XP_011519396.1 | 4, 5 | 618 | |||
XM_011521095.1 | XP_011519397.1 | 4, 5 | 618 | |||
XM_011521096.1 | XP_011519398.1 | 4, 5 | 618 | |||
XM_011521097.1 | XP_011519399.1 | 4, 5 | 618 | |||
XM_011521100.2 | XP_011519402.1 | 4, 5 | 618 | |||
XM_017020662.1 | XP_016876151.1 | 4, 5 | 618 | |||
XM_017020663.1 | XP_016876152.1 | 4, 5 | 582 | |||
GenBank mRNA | AF227906.2 | - | 4, 5 | 564 | ||
AK001735.1 | - | 4, 5 | 588 | |||
AL133051.1 | - | 1, 2 | 43 | |||
BC017759.1 | - | 4, 5 | 567 | |||
BC032302.1 | - | 4, 5 | 663 | |||
BC067807.1 | - | 4, 5 | 574 | |||
BC111021.1 | - | 4, 5 | 581 | |||
BC125233.1 | - | 4, 5 | 519 | 40124676 | ||
BC125234.1 | - | 4, 5 | 519 |