Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020123.3 | NP_064508.3 | 4 | 660 | ||
XM_011539976.2 | XP_011538278.1 | 4 | 740 | |||
XM_011539977.1 | XP_011538279.1 | 4 | 740 | |||
GenBank mRNA | AF116347.1 | - | 4 | 410 | ||
AF160213.1 | - | 4 | 459 | |||
AF269150.1 | - | 4 | 660 | |||
AK000756.1 | - | 2 | 63 | |||
AK027788.1 | - | 3 | 316 | |||
AK314348.1 | - | 4 | 626 | |||
AY358819.1 | - | 3 | 330 | |||
BC020959.1 | - | 3 | 240 | |||
BC136788.1 | - | 4 | 660 | 9020778 | ||
BC136789.1 | - | 4 | 660 | 9020779 |