Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020137.4 | NP_064522.3 | 15 | 1279 | ||
XM_011543935.1 | XP_011542237.1 | 15 | 1279 | |||
XM_011543936.1 | XP_011542238.1 | 15 | 1279 | |||
XM_011543937.1 | XP_011542239.1 | 15 | 1326 | |||
XM_017029656.1 | XP_016885145.1 | 15 | 1279 | |||
XM_017029657.1 | XP_016885146.1 | 15 | 1650 | |||
XM_017029658.1 | XP_016885147.1 | 15 | 2589 | |||
XM_017029659.1 | XP_016885148.1 | 15 | 1326 | |||
XM_017029660.1 | XP_016885149.1 | 15 | 1680 | |||
XM_017029661.1 | XP_016885150.1 | 15 | 1326 | |||
GenBank mRNA | AB032993.1 | - | 15 | 1224 | ||
AK292158.1 | - | 15 | 1250 | |||
AL136847.1 | - | 2 | 165 | |||
BC001522.2 | - | 5 | 392 | |||
BC101544.1 | - | 15 | 1277 | 8069050 | ||
BC101546.1 | - | 15 | 1277 | 8069052 | ||
BX640704.1 | - | 9 | 963 | |||
BX647801.1 | - | 14 | 1119 | |||
BX647804.1 | - | 14 | 1119 |