Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam58b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025412.1 | NP_001020583.1 | 1 | 400 | ||
GenBank mRNA | BC086445.1 | - | 1 | 387 | ||
BC097941.1 | - | 1 | 400 | 7376750 |