Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025700.1 | NP_001020871.1 | 2 | 1322 | ||
XM_006242639.2 | XP_006242701.1 | 3 | 1685 | |||
GenBank mRNA | BC098684.1 | - | 2 | 1322 | 7385284 |