Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025707.1 | NP_001020878.1 | 1 | 175 | ||
GenBank mRNA | BC099102.1 | - | 1 | 175 | 7460021 |