Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025711.1 | NP_001020882.1 | 7 | 1792 | ||
XM_006245533.2 | XP_006245595.1 | 13 | 2332 | |||
XM_006245534.2 | XP_006245596.1 | 12 | 2235 | |||
GenBank mRNA | BC098833.1 | - | 7 | 1792 | 7378703 |