Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199699.1 | NP_001186628.1 | 2 | 695 | ||
NM_020162.3 | NP_064547.2 | 3 | 856 | |||
GenBank mRNA | AK025625.1 | - | 3 | 657 | ||
AK295074.1 | - | 2 | 464 | |||
AK295760.1 | - | 3 | 856 | |||
AK314580.1 | - | 3 | 824 | |||
BC030017.1 | - | 2 | 382 |