Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025717.1 | NP_001020888.1 | 4 | 882 | ||
GenBank mRNA | BC099238.1 | - | 4 | 882 | 7445509 | |
FQ225700.1 | - | 2 | 506 | |||
FQ230401.1 | - | 2 | 506 | |||
FQ230694.1 | - | 1 | 102 |