Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025722.2 | NP_001020893.2 | 3, 4 | 791 | ||
NM_001109936.1 | NP_001103406.1 | 4, 5 | 340 | |||
NM_001109937.1 | NP_001103407.1 | 4, 5 | 301 | |||
GenBank mRNA | BC098804.1 | - | 3, 4 | 791 | 7376321 | |
DN932869.1 | - | 4, 5 | 336 | |||
DN935585.1 | - | 4, 5 | 360 | |||
DY320517.1 | - | 4, 5 | 341 |