Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321463.1 | NP_001308392.1 | 7 | 994 | ||
NM_020170.3 | NP_064555.2 | 7 | 994 | |||
GenBank mRNA | AK125002.1 | - | 7 | 1050 | ||
AK172848.1 | - | 7 | 941 | |||
AK314044.1 | - | 7 | 929 | |||
AL136567.1 | - | 7 | 825 | |||
BC008920.2 | - | 3 | 201 | |||
BC010064.2 | - | 7 | 910 | 4100399 | ||
BC013283.2 | - | 7 | 1271 | 3959506 | ||
BC019091.2 | - | 7 | 910 | 4895124 | ||
BC025926.1 | - | 7 | 906 | 3509966 | ||
CR533566.1 | - | 7 | 807 |