Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Procr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025733.2 | NP_001020904.1 | 4 | 1213 | ||
GenBank mRNA | BC097380.1 | - | 4 | 1184 | 7445753 | |
BC099115.1 | - | 4 | 1213 | 7462042 | ||
FQ220150.1 | - | 4 | 1252 | |||
FQ221148.1 | - | 4 | 1223 | |||
FQ228484.1 | - | 4 | 1218 | |||
FQ228710.1 | - | 4 | 1220 | |||
FQ229483.1 | - | 4 | 1221 |