Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082533.1 | NP_001076002.1 | 4, 5 | 1399 | ||
NM_001082534.1 | NP_001076003.1 | 4, 5 | 964 | |||
NM_020178.4 | NP_064563.1 | 3, 4 | 1273 | |||
GenBank mRNA | AB001597.1 | - | 3, 4 | 287 | ||
AB036836.1 | - | 4, 5 | 1189 | |||
AF288385.1 | - | 4, 5 | 955 | |||
AK294652.1 | - | 2, 3 | 644 | |||
AK313001.1 | - | 3, 4 | 849 | |||
AL161812.1 | - | 3, 4 | 289 | |||
AY358509.1 | - | 3, 4 | 567 | |||
BC020577.1 | - | 4, 5 | 931 | 4181377 | ||
BC029865.1 | - | 4, 5 | 933 | |||
BC047456.1 | - | 3, 4 | 1186 | 5276935 | ||
BC068462.1 | - | 3, 4 | 1232 | 5260321 | ||
CR749607.1 | - | 4, 5 | 735 |