Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025748.1 | NP_001020919.1 | 10 | 1726 | ||
XM_006229599.2 | XP_006229661.1 | 11 | 2117 | |||
XM_006229600.2 | XP_006229662.1 | 12 | 1908 | |||
GenBank mRNA | BC099221.1 | - | 10 | 1726 | 7387882 |