Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF4 SIRNA Assays ›
Gene Symbol:
CELF4
Gene Name:
CUGBP, Elav-like family member 4
Gene Aliases:
BRUNOL4, CELF-4
Chromosome Location:
Chr.18: 37243040 - 37566052 on Build GRCh38
UniGene ID:
Hs.435976
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28117
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025087.1
NP_001020258.1
1
665
NM_001025088.1
NP_001020259.1
1
665
NM_001025089.1
NP_001020260.1
1
665
NM_020180.3
NP_064565.1
1
665
XM_005258308.3
XP_005258365.1
1
677
XM_011526079.2
XP_011524381.1
1
679
XM_011526081.2
XP_011524383.1
1
516
XM_011526083.2
XP_011524385.1
1
679
XM_011526084.2
XP_011524386.1
1
679
XM_011526085.2
XP_011524387.1
1
680
XM_011526086.2
XP_011524388.1
1
678
XM_011526087.2
XP_011524389.1
1
680
XM_011526090.2
XP_011524392.1
1
677
XM_011526091.2
XP_011524393.1
1
677
XM_011526092.2
XP_011524394.1
1
677
XM_017025840.1
XP_016881329.1
1
542
XM_017025841.1
XP_016881330.1
1
676
XM_017025842.1
XP_016881331.1
1
678
XM_017025843.1
XP_016881332.1
1
678
XM_017025844.1
XP_016881333.1
1
678
XM_017025845.1
XP_016881334.1
1
678
XM_017025846.1
XP_016881335.1
1
676
XM_017025847.1
XP_016881336.1
1
676
XM_017025848.1
XP_016881337.1
1
677
XM_017025849.1
XP_016881338.1
1
677
XM_017025850.1
XP_016881339.1
1
542
XM_017025851.1
XP_016881340.1
1
542
XM_017025852.1
XP_016881341.1
1
542
XM_017025853.1
XP_016881342.1
1
678
XM_017025854.1
XP_016881343.1
1
678
XM_017025855.1
XP_016881344.1
1
678
XM_017025856.1
XP_016881345.1
1
678
XM_017025857.1
XP_016881346.1
1
679
XM_017025858.1
XP_016881347.1
1
516
XM_017025859.1
XP_016881348.1
1
516
XM_017025860.1
XP_016881349.1
1
677
XM_017025861.1
XP_016881350.1
1
542
XM_017025862.1
XP_016881351.1
1
542
XM_017025863.1
XP_016881352.1
1
524
XM_017025864.1
XP_016881353.1
1
516
XM_017025865.1
XP_016881354.1
1
542
GenBank mRNA
AB209774.1
-
1
459
AF248650.1
-
1
404
AF329265.1
-
1
269
AK074596.1
-
1
397
AK295009.1
-
1
279
BC001946.1
-
1
426
2820541
BC004167.2
-
1
426
2820541
BC045711.1
-
1
352
5261155
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107400.This siRNA also targets the following transcripts in Mouse: NM_001174074, NM_133195, NM_001146294, NM_001146292, NM_001146295, and others.
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
germ cell development
embryo development
negative regulation of translation
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA splicing, via spliceosome
negative regulation of excitatory postsynaptic potential
nucleotide binding
translation repressor activity, nucleic acid binding
mRNA binding
BRE binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0