Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037158.1 | NP_001032235.1 | 1 | 595 | ||
GenBank mRNA | AY574020.1 | - | 1 | 595 |