Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031637.1 | NP_001026807.1 | 22 | 3703 | ||
XM_006245927.2 | XP_006245989.1 | 23 | 4811 | |||
XM_006245928.2 | XP_006245990.1 | 23 | 4795 | |||
XM_008767554.1 | XP_008765776.1 | 23 | 5425 | |||
XM_008767555.1 | XP_008765777.1 | 22 | 3798 | |||
XM_008767556.1 | XP_008765778.1 | 22 | 3684 | |||
XM_008767557.1 | XP_008765779.1 | 22 | 3717 | |||
GenBank mRNA | AB196699.1 | - | 22 | 3703 |