Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033685.2 | NP_001028857.2 | 3 | 262 | ||
GenBank mRNA | BC104706.1 | - | 3 | 251 | 7316605 | |
CK357541.1 | - | 3 | 219 | |||
FQ212380.1 | - | 3 | 234 | |||
FQ223271.1 | - | 3 | 229 | |||
FQ234134.1 | - | 3 | 270 |
Gene Symbol: RGD1561590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727811.4 | XP_002727857.1 | 1 | 285 |