Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033690.1 | NP_001028862.1 | 3 | 530 | ||
XM_006255357.1 | XP_006255419.1 | 2 | 120 | |||
GenBank mRNA | BC101871.1 | - | 3 | 530 | 5621693 |