Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037094.1 | NP_001032171.1 | 3 | 176 | ||
GenBank mRNA | BC079393.1 | - | 3 | 226 | ||
BC089862.1 | - | 3 | 174 | |||
BC107464.1 | - | 3 | 176 | 7454431 |