Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cblc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034920.2 | NP_001030092.1 | 6 | 1014 | ||
GenBank mRNA | BC105776.1 | - | 6 | 999 | 7931977 |