Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4ENIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164501.1 | NP_001157973.1 | 19 | 3460 | ||
NM_001164502.1 | NP_001157974.1 | 17 | 2962 | |||
NM_019843.3 | NP_062817.2 | 19 | 3484 | |||
XM_005261686.2 | XP_005261743.1 | 19 | 3487 | |||
XM_005261687.2 | XP_005261744.1 | 19 | 3468 | |||
XM_005261688.2 | XP_005261745.1 | 19 | 3338 | |||
XM_006724281.3 | XP_006724344.3 | 19 | 3764 | |||
XM_011530280.1 | XP_011528582.1 | 19 | 3335 | |||
XM_011530281.2 | XP_011528583.1 | 19 | 3559 | |||
XM_017028863.1 | XP_016884352.1 | 19 | 3761 | |||
XM_017028864.1 | XP_016884353.1 | 19 | 3556 | |||
GenBank mRNA | AF240775.1 | - | 19 | 3313 | ||
AK025254.1 | - | 12 | 2215 | |||
AK074768.1 | - | 5 | 1259 | |||
AK130061.1 | - | 9 | 2655 | |||
BC032941.1 | - | 17 | 2903 | 5268671 | ||
BC033028.1 | - | 19 | 3434 | 4837225 |