Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037772.1 | NP_001032861.2 | 8, 9 | 1057 | ||
NM_001244805.1 | NP_001231734.1 | 8, 9 | 1226 | |||
XM_008766490.1 | XP_008764712.1 | 10, 11 | 1348 | |||
XM_008766491.1 | XP_008764713.1 | 9, 10 | 1072 | |||
XM_008766492.1 | XP_008764714.1 | 8, 9 | 2647 | |||
XM_008766493.1 | XP_008764715.1 | 10, 11 | 1223 | |||
GenBank mRNA | BC083632.1 | - | 11, 12 | 1310 | ||
BC098948.1 | - | 8, 9 | 1057 | |||
BC161815.1 | - | 8, 9 | 1226 | 9031816 |