Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033893.1 | NP_001029065.1 | 4 | 1541 | ||
XM_008767681.1 | XP_008765903.1 | 1764 | ||||
GenBank mRNA | DQ176639.1 | - | 4 | 1541 |
Gene Symbol: LOC103689949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246253.2 | XP_006246315.1 | 5 | 1764 |