Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034143.1 | NP_001029315.1 | 3 | 373 | ||
GenBank mRNA | BC105774.1 | - | 3 | 373 | 7930977 |