Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020133.2 | NP_064518.1 | 9 | 1536 | ||
XM_005267053.3 | XP_005267110.1 | 8 | 1352 | |||
XM_006715514.3 | XP_006715577.1 | 8 | 1360 | |||
XM_017011059.1 | XP_016866548.1 | 10 | 1706 | |||
GenBank mRNA | AF156776.1 | - | 9 | 1466 | ||
AJ420473.1 | - | 9 | 1508 | |||
AK074208.1 | - | 9 | 1522 | |||
AK299721.1 | - | 6 | 1020 | |||
AL832128.1 | - | 1 | 5249 | |||
AY358506.1 | - | 9 | 1476 | |||
BC013410.2 | - | 2 | 396 | |||
BC020209.1 | - | 9 | 1468 | 4850399 | ||
CR627395.1 | - | 8 | 1317 |