Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033697.1 | NP_001028869.1 | 9 | 1101 | ||
XM_006254188.2 | XP_006254250.1 | 8 | 1072 | |||
GenBank mRNA | BC088227.1 | - | 9 | 1090 | ||
BC104716.1 | - | 9 | 1101 | 7373256 |