Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034187.1 | NP_001029359.1 | 8 | 1960 | ||
GenBank mRNA | BC100155.1 | - | 8 | 1960 | 7461488 |