Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037646.1 | NP_001032735.1 | 2 | 351 | ||
XM_006229059.2 | XP_006229121.1 | 2 | 390 | |||
XM_006229060.2 | XP_006229122.1 | 2 | 390 | |||
XM_006229061.2 | XP_006229123.1 | 2 | 391 | |||
XM_008759373.1 | XP_008757595.1 | 2 | 391 | |||
XM_008759374.1 | XP_008757596.1 | 2 | 391 | |||
GenBank mRNA | BC107809.1 | - | 2 | 351 | 7453736 | |
FQ228777.1 | - | 2 | 336 |