Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015028.2 | NP_001015028.2 | 16 | 3988 | ||
GenBank mRNA | AY661303.1 | - | 16 | 3988 | ||
BC091436.1 | - | 2 | 1250 |