Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761975.1 | XP_008760197.1 | 4 | 729 | ||
GenBank mRNA | AA875516.1 | - | 2 | 231 |