Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPAN-P2RY11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040664.2 | NP_001035754.1 | 2, 3 | 369 | ||
NM_001198690.1 | NP_001185619.1 | 2, 3 | 369 | |||
GenBank mRNA | AJ300588.1 | - | 2, 3 | 197 | ||
BC171852.1 | - | 2, 3 | 369 |
Gene Symbol: PPAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020230.5 | NP_064615.3 | 2, 3 | 369 | ||
GenBank mRNA | AJ292529.1 | - | 2, 3 | 224 | ||
BC009833.2 | - | 2, 3 | 241 | 3957286 | ||
BC033202.1 | - | 2, 3 | 303 | 4871446 |