Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034032.1 | NP_001029204.1 | 3 | 226 | ||
GenBank mRNA | AB062135.1 | - | 3 | 228 | ||
BC100072.1 | - | 3 | 226 | 7325259 | ||
FQ214972.1 | - | 3 | 239 | |||
FQ219445.1 | - | 3 | 239 |