Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Popdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037369.1 | NP_001032446.1 | 2 | 486 | ||
XM_006256599.2 | XP_006256661.1 | 2 | 567 | |||
GenBank mRNA | AY787642.1 | - | 1 | 294 | ||
BC105761.1 | - | 2 | 486 | 5623862 | ||
FQ218039.1 | - | 2 | 388 |