Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135823.1 | NP_001129295.1 | 2 | 255 | ||
NM_005086.4 | NP_005077.2 | 2 | 438 | |||
XM_011520853.2 | XP_011519155.1 | 2 | 188 | |||
XM_011520854.2 | XP_011519156.1 | 2 | 362 | |||
XM_011520855.2 | XP_011519157.1 | 2 | 104 | |||
GenBank mRNA | AF016028.2 | - | 2 | 338 | ||
AK092925.1 | - | 2 | 251 | |||
AK304667.1 | - | 3 | 332 | |||
AK314353.1 | - | 2 | 427 | |||
AL136756.1 | - | 2 | 422 | |||
BC062299.1 | - | 2 | 418 | 6376868 | ||
BP341300.1 | - | 2 | 438 | |||
DA073350.1 | - | 2 | 255 | |||
EU433933.1 | - | 2 | 338 | |||
X89105.1 | - | 2 | 260 |