Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026166.2 | NP_080442.2 | 2 | 300 | ||
NM_027078.2 | NP_081354.1 | 2 | 300 | |||
GenBank mRNA | AK004670.1 | - | 2 | 258 | ||
AK006283.1 | - | 2 | 268 | |||
BC034890.1 | - | 2 | 262 | 1447362 | ||
BC055834.1 | - | 2 | 251 | 3492302 | ||
CF171082.1 | - | 2 | 282 |