Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025559.3 | NP_001020730.1 | 16 | 6961 | ||
NM_001025560.2 | NP_001020731.1 | 16 | 7075 | |||
NM_001277326.1 | NP_001264255.1 | 16 | 7056 | |||
NM_001277327.1 | NP_001264256.1 | 15 | 7116 | |||
NM_001277328.1 | NP_001264257.1 | 14 | 6821 | |||
NM_011445.4 | NP_035575.1 | 17 | 7109 | |||
GenBank mRNA | AK030232.1 | - | 1 | 875 | ||
AK044981.1 | - | 1 | 2313 |