Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011296.2 | NP_035426.1 | 3 | 293 | ||
GenBank mRNA | AK050626.1 | - | 3 | 316 | ||
AK146354.1 | - | 3 | 316 | |||
AK151580.1 | - | 3 | 316 | |||
AK159437.1 | - | 3 | 316 | |||
AK159821.1 | - | 3 | 316 | |||
AK169246.1 | - | 3 | 316 | |||
AK169732.1 | - | 2 | 937 | |||
BC081458.1 | - | 2 | 281 | 5712669 | ||
BC081459.1 | - | 3 | 293 | 6407328 | ||
BC091729.1 | - | 3 | 291 | |||
BC093500.1 | - | 3 | 294 | 4207659 | ||
BC100604.1 | - | 3 | 315 | 30120583 | ||
M76763.1 | - | 3 | 294 |