Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011056.3 | NP_035186.1 | 17 | 2189 | ||
GenBank mRNA | AF536978.1 | - | 17 | 2189 | ||
AK081466.1 | - | 10 | 1562 | |||
AY388962.1 | - | 15 | 1802 | |||
BC132107.1 | - | 16 | 2057 | 40130384 | ||
BC132109.1 | - | 16 | 2057 | 40130386 | ||
DQ665891.1 | - | 10 | 1659 | |||
DQ665892.1 | - | 10 | 1445 | |||
DQ665893.1 | - | 11 | 1573 | |||
DQ665894.1 | - | 11 | 1417 | |||
DQ665895.1 | - | 10 | 1700 | |||
DQ665896.1 | - | 10 | 1489 | |||
EU489880.1 | - | 15 | 2045 |