Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020358.2 | NP_065091.1 | 5 | 861 | ||
GenBank mRNA | AB037682.1 | - | 5 | 861 | ||
BC075019.2 | - | 4 | 560 | 30915409 | ||
BC075020.2 | - | 4 | 560 | 30915620 | ||
BC126472.1 | - | 4 | 655 | 8992188 | ||
JF968461.1 | - | 3 | 292 |
Gene Symbol: TRIM49B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206626.1 | NP_001193555.1 | 3 | 532 | ||
GenBank mRNA | JF968463.1 | - | 3 | 418 |
Gene Symbol: TRIM49C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195234.1 | NP_001182163.1 | 5 | 861 | ||
GenBank mRNA | BC126470.1 | - | 4 | 655 | 8992186 | |
JF968462.1 | - | 3 | 292 |
Gene Symbol: TRIM53CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JF968458.1 | - | 3 | 250 |