Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009697.3 | NP_033827.2 | 3 | 1561 | ||
NM_183261.3 | NP_899084.2 | 3 | 1509 | |||
GenBank mRNA | AK032882.1 | - | 1 | 633 | ||
AK034619.1 | - | 2 | 680 | |||
AK140125.1 | - | 3 | 1485 | |||
BC042484.1 | - | 3 | 1561 | 5368085 | ||
BC094360.1 | - | 3 | 1486 | |||
U07635.1 | - | 3 | 1444 |