Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286521.1 | NP_001273450.1 | 5, 6 | 654 | ||
NM_001286522.1 | NP_001273451.1 | 6, 7 | 776 | |||
NM_020367.5 | NP_065100.2 | 6, 7 | 775 | |||
XM_005253711.4 | XP_005253768.1 | 6, 7 | 1169 | |||
XM_011520970.1 | XP_011519272.1 | 7, 8 | 891 | |||
XM_011520971.2 | XP_011519273.1 | 7, 8 | 1291 | |||
XM_011520973.2 | XP_011519275.1 | 6, 7 | 604 | |||
XM_017019670.1 | XP_016875159.1 | 7, 8 | 813 | |||
XM_017019671.1 | XP_016875160.1 | 6, 7 | 1170 | |||
GenBank mRNA | AF263540.1 | - | 5, 6 | 590 | ||
AK299372.1 | - | 5, 6 | 507 | |||
BC017569.1 | - | 6, 7 | 701 | 4077842 | ||
BC031073.1 | - | 6, 7 | 763 | 5284275 | ||
BX640960.1 | - | 5, 6 | 578 | |||
BX647487.1 | - | 6, 7 | 694 | |||
CR749294.1 | - | 6, 7 | 682 |