Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020368.2 | NP_065101.1 | 1 | 348 | ||
GenBank mRNA | AF271212.1 | - | 1 | 310 | ||
AK026909.1 | - | 1 | 297 | |||
AL136590.1 | - | 1 | 283 | |||
BC004546.1 | - | 1 | 290 | 3946633 | ||
CR533544.1 | - | 1 | 149 | |||
DB021011.1 | - | 1 | 348 |