Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133766.3 | NP_598527.2 | 6 | 715 | ||
GenBank mRNA | AB158474.1 | - | 6 | 719 | ||
AK030074.1 | - | 6 | 716 | |||
AK032461.1 | - | 6 | 688 | |||
AK044000.1 | - | 6 | 709 | |||
AK044475.1 | - | 6 | 711 | |||
AK129065.1 | - | 6 | 693 | |||
AK161250.1 | - | 6 | 700 | |||
BC007482.1 | - | 6 | 719 | 3495608 | ||
BC032092.1 | - | 6 | 720 |