Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1b27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009596.1 | NP_033726.1 | 2 | 218 | ||
GenBank mRNA | AF039064.1 | - | 1 | 151 | ||
BC016132.1 | - | 2 | 218 | 4922791 | ||
BC132436.1 | - | 2 | 302 | 40130713 | ||
GU269231.1 | - | 2 | 217 |