Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020410.2 | NP_065143.2 | 3 | 540 | ||
GenBank mRNA | AB058728.1 | - | 3 | 510 | ||
AF288687.1 | - | 3 | 158 | |||
AK056420.1 | - | 3 | 516 |